Assay Details
Target Gene Details
Entrez Gene ID: | 7411 |
Gene Name: | VHL binding protein 1 |
Gene Aliases: |
HIBBJ46, PFD3, PFDN3, VBP-1 |
Location: |
Chr.X:155197007-155239841 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VBP1 | NM_001303543.1 | NP_001290472.1 | ||
| NM_001303544.1 | NP_001290473.1 | |||
| NM_001303545.1 | NP_001290474.1 | |||
| NM_003372.6 | NP_003363.1 | |||
| AK301646.1 | ||||
| AK313420.1 | ||||
| BC046094.1 | AAH46094.1 | |||
| BT019604.1 | AAV38411.1 | |||
| CR541932.1 | CAG46730.1 | |||
| DA610631.1 | ||||
| U56833.1 | AAC50617.1 | |||
| U96759.1 | AAC23907.1 | |||
| Y17394.1 | CAA76761.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| esv3575068 | Chr.X:155215605 - 155223004 on Build GRCh38 | Loss |
|
| esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map