Assay Details
Target Gene Details
Entrez Gene ID: | 5394 |
Gene Name: | exosome component 10 |
Gene Aliases: |
PM-Scl, PM/Scl-100, PMSCL, PMSCL2, RRP6, Rrp6p, p2, p3, p4 |
Location: |
Chr.1:11066613-11099910 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOSC10 | NM_001001998.2 | NP_001001998.1 | ||
| NM_002685.3 | NP_002676.1 | |||
| XM_005263475.2 | XP_005263532.2 | |||
| XM_017001494.1 | XP_016856983.1 | |||
| XM_017001495.1 | XP_016856984.1 | |||
| XM_017001496.1 | XP_016856985.1 | |||
| XM_017001497.1 | XP_016856986.1 | |||
| AB209236.1 | BAD92473.1 | |||
| AK123365.1 | ||||
| AK296559.1 | ||||
| BC009908.2 | AAH09908.2 | |||
| BC039901.1 | AAH39901.1 | |||
| BC073788.1 | AAH73788.1 | |||
| CA307576.1 | ||||
| L01457.1 | AAB59352.1 | |||
| X66113.1 | CAA46904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545407 | Chr.1:11000018 - 11377969 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map