Assay Details
Target Gene Details
Entrez Gene ID: | 65108 |
Gene Name: | MARCKS like 1 |
Gene Aliases: |
F52, MACMARCKS, MLP, MLP1, MRP |
Location: |
Chr.1:32333829-32336239 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARCKSL1 | NM_023009.6 | NP_075385.1 | ||
| NR_052852.1 | ||||
| AI879076.1 | ||||
| AL713653.1 | CAD28462.1 | |||
| AY007131.1 | ||||
| BC007904.2 | AAH07904.1 | |||
| BC066915.1 | AAH66915.1 | |||
| DT218825.1 | ||||
| X70326.1 | CAA49793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1112529 | Chr.1:32335399 - 32336599 on Build GRCh38 | Deletion |
|
| nsv1077308 | Chr.1:32335398 - 32335799 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map