Assay Details
Target Gene Details
Entrez Gene ID: | 163259 |
Gene Name: | DENN domain containing 2C |
Gene Aliases: |
dJ1156J9.1 |
Location: |
Chr.1:114584575-114670111 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND2C | NM_001256404.1 | NP_001243333.1 | ||
| NM_198459.3 | NP_940861.3 | |||
| BX647950.1 | ||||
| BX649075.1 | ||||
| CR749576.1 | CAH18369.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28734 | Chr.1:114376708 - 115005298 on Build GRCh38 | Loss |
|
| nsv831104 | Chr.1:114455501 - 114627235 on Build GRCh38 | Loss |
|
| esv3587266 | Chr.1:114587471 - 114614453 on Build GRCh38 | Loss |
|
| esv3587267 | Chr.1:114592824 - 114620609 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map