Assay Details
Target Gene Details
Entrez Gene ID: | 374986 |
Gene Name: | mitoguardin 1 |
Gene Aliases: |
FAM73A |
Location: |
Chr.1:77779624-77879540 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIGA1 | NM_001270384.1 | NP_001257313.1 | ||
| NM_198549.3 | NP_940951.1 | |||
| XM_005270846.4 | XP_005270903.1 | |||
| XM_005270847.4 | XP_005270904.1 | |||
| XM_005270848.4 | XP_005270905.1 | |||
| XM_011541422.2 | XP_011539724.1 | |||
| XM_017001227.1 | XP_016856716.1 | |||
| XM_017001228.1 | XP_016856717.1 | |||
| XM_017001229.1 | XP_016856718.1 | |||
| XM_017001230.1 | XP_016856719.1 | |||
| XM_017001231.1 | XP_016856720.1 | |||
| XM_017001232.1 | XP_016856721.1 | |||
| AK092412.1 | BAC03880.1 | |||
| AK296411.1 | ||||
| BC130387.1 | ||||
| BC144167.1 | ||||
| BX537792.1 | CAD97844.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546591 | Chr.1:77308728 - 77803522 on Build GRCh38 | Gain |
|
| nsv830303 | Chr.1:77625139 - 77843080 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map