Assay Details
Target Gene Details
Entrez Gene ID: | 256435 |
Gene Name: | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 3 |
Gene Aliases: |
PRO7177, SIAT7C, ST6GALNACIII, STY |
Location: |
Chr.1:76074684-76632880 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GALNAC3 | NM_001160011.1 | NP_001153483.1 | ||
| NM_152996.2 | NP_694541.2 | |||
| XM_006710553.3 | XP_006710616.1 | |||
| XM_017000940.1 | XP_016856429.1 | |||
| AJ507291.1 | CAD45371.1 | |||
| AK091215.1 | BAC03611.1 | |||
| AK297363.1 | ||||
| AY358540.1 | AAQ88904.1 | |||
| BC059363.1 | AAH59363.1 | |||
| DB473741.1 |
More Information
Set Membership: |
|

Genomic Map