Assay Details
Target Gene Details
Entrez Gene ID: | 101929093 |
Gene Name: | long intergenic non-protein coding RNA 1350 |
Gene Aliases: |
GS1-204I12.1 |
Location: |
Chr.1:185558379-185628488 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01350 | NR_110793.1 | |||
| AF508906.1 | ||||
| AF510427.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548398 | Chr.1:185167326 - 186033962 on Build GRCh38 | Gain |
|

Genomic Map