Assay Details
Target Gene Details
Entrez Gene ID: | 5825 |
Gene Name: | ATP binding cassette subfamily D member 3 |
Gene Aliases: |
ABC43, CBAS5, PMP70, PXMP1, ZWS2 |
Location: |
Chr.1:94385106-94518663 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCD3 | XM_017001953.1 | XP_016857442.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv946085 | Chr.1:94401746 - 94408931 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map