Assay Details
Target Gene Details
Entrez Gene ID: | 2140 |
Gene Name: | EYA transcriptional coactivator and phosphatase 3 |
Gene Aliases: |
- |
Location: |
Chr.1:27970344-28088671 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EYA3 | NM_001282561.1 | 17 | 5393 | NP_001269490.1 |
| NM_001282562.1 | 17 | 5451 | NP_001269491.1 | |
| NM_001990.3 | 18 | 5531 | NP_001981.2 | |
| XM_006710449.3 | 19 | 5567 | XP_006710512.1 | |
| XM_011541004.2 | 18 | 5427 | XP_011539306.1 | |
| BX648945.1 | 18 | 5500 | ||
| CA434977.1 | 1 | 511 |
More Information
Set Membership: |
|

Genomic Map