Assay Details
Target Gene Details
Entrez Gene ID: | 284576 |
Gene Name: | LEMD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:205373252-205387440 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LEMD1-AS1 | NR_038425.1 | |||
AK090497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832381 | Chr.1:205290241 - 205479588 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1014629 | Chr.1:205367415 - 205435109 on Build GRCh38 | Gain |
![]() ![]() ![]() |
