Assay Details
Target Gene Details
Entrez Gene ID: | 255104 |
Gene Name: | transmembrane and coiled-coil domains 4 |
Gene Aliases: |
- |
Location: |
Chr.1:19682213-19800385 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508991 | Chr.1:19567574 - 19705911 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75316091] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map