Assay Details
Target Gene Details
Entrez Gene ID: | 10905 |
Gene Name: | mannosidase alpha class 1A member 2 |
Gene Aliases: |
MAN1B |
Location: |
Chr.1:117367393-117528872 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAN1A2 | NM_006699.3 | NP_006690.1 | ||
| XM_006710302.2 | XP_006710365.1 | |||
| XM_011540536.2 | XP_011538838.1 | |||
| XM_017000115.1 | XP_016855604.1 | |||
| AF027156.1 | AAC26169.1 | |||
| AK307842.1 | ||||
| AU133800.1 | ||||
| BC063300.1 | AAH63300.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009706 | Chr.1:117049372 - 117442154 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map