Assay Details
Target Gene Details
Entrez Gene ID: | 400799 |
Gene Name: | long intergenic non-protein coding RNA 1344 |
Gene Aliases: |
GS1-122H1.2 |
Location: |
Chr.1:182203945-182314061 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01344 | NR_104175.1 | |||
| AK128015.1 | ||||
| BX096536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|

Genomic Map