Assay Details
Target Gene Details
Entrez Gene ID: | 23341 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C16 |
Gene Aliases: |
- |
Location: |
Chr.1:15526694-15571733 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC16 | NM_001287811.1 | NP_001274740.1 | ||
| NM_015291.3 | NP_056106.1 | |||
| NR_109898.1 | ||||
| AB023179.2 | ||||
| AK023805.1 | ||||
| AK091848.1 | ||||
| AK292479.1 | ||||
| AL833299.1 | ||||
| BC047363.1 | AAH47363.1 | |||
| BX648982.1 | ||||
| CR749259.1 | CAH18115.1 | |||
| CR749567.1 | CAH18363.1 |
More Information
Set Membership: |
|

Genomic Map