Assay Details
Target Gene Details
Entrez Gene ID: | 2275 |
Gene Name: | four and a half LIM domains 3 |
Gene Aliases: |
SLIM2 |
Location: |
Chr.1:37996765-38005515 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FHL3 | NM_001243878.1 | NP_001230807.1 | ||
| NM_004468.4 | NP_004459.2 | |||
| AK290641.1 | ||||
| AK309179.1 | ||||
| BC001351.2 | AAH01351.1 | |||
| BC011697.2 | AAH11697.1 | |||
| BC014501.1 | AAH14501.1 | |||
| BX384389.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950632 | Chr.1:38004129 - 38006228 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map