Assay Details
Target Gene Details
Entrez Gene ID: | 104355292 |
Gene Name: | long intergenic non-protein coding RNA 1525 |
Gene Aliases: |
lnc-MAN1A2-1 |
Location: |
Chr.1:117295466-117321336 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01525 | NR_126408.1 | |||
| BX109912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009706 | Chr.1:117049372 - 117442154 on Build GRCh38 | Gain |
|

Genomic Map