Assay Details
Target Gene Details
Entrez Gene ID: | 5293 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta |
Gene Aliases: |
APDS, IMD14, P110DELTA, PI3K, p110D |
Location: |
Chr.1:9629889-9729114 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3CD | NM_005026.3 | NP_005017.3 | ||
| XM_006710689.2 | XP_006710752.1 | |||
| XM_017001477.1 | XP_016856966.1 | |||
| XM_017001478.1 | XP_016856967.1 | |||
| XM_017001479.1 | XP_016856968.1 | |||
| XM_017001480.1 | XP_016856969.1 | |||
| XM_017001481.1 | XP_016856970.1 | |||
| AK314191.1 | ||||
| BC150297.1 | ||||
| CN313981.1 | ||||
| U86453.1 | AAC25677.1 | |||
| Y10055.2 | CAA71149.2 |
Target Gene Details
Entrez Gene ID: | 101929074 |
Gene Name: | PIK3CD antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.1:9672426-9687574 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3CD-AS2 | NR_126366.1 | |||
| NR_126367.1 | ||||
| NR_126368.1 | ||||
| AA341665.1 | ||||
| AL523099.3 | ||||
| BI763597.1 | ||||
| BP394863.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585185 | Chr.1:9670031 - 9692162 on Build GRCh38 | Gain |
|
| nsv832203 | Chr.1:9666574 - 9800151 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115045980] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map