Assay Details
Target Gene Details
Entrez Gene ID: | 100506963 |
Gene Name: | TCEB3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:23760382-23778297 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCEB3-AS1 | NR_038280.1 | |||
| AL049675.1 | ||||
| BG429664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv29e199 | Chr.1:23764964 - 23773446 on Build GRCh38 | Deletion |
|
| esv3585491 | Chr.1:23764803 - 23773585 on Build GRCh38 | Loss |
|

Genomic Map