Assay Details
Target Gene Details
Entrez Gene ID: | 142 |
Gene Name: | poly(ADP-ribose) polymerase 1 |
Gene Aliases: |
ADPRT, ADPRT 1, ADPRT1, ARTD1, PARP, PARP-1, PPOL, pADPRT-1 |
Location: |
Chr.1:226360691-226408100 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP1 | NM_001618.3 | NP_001609.2 | ||
| AK225654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832770 | Chr.1:226277429 - 226467811 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map