Assay Details
Target Gene Details
Entrez Gene ID: | 10236 |
Gene Name: | heterogeneous nuclear ribonucleoprotein R |
Gene Aliases: |
HNRPR, hnRNP-R |
Location: |
Chr.1:23304688-23344650 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPR | NM_001102397.2 | 10 | 5851 | NP_001095867.1 |
| NM_001102398.2 | 11 | 6026 | NP_001095868.1 | |
| NM_001102399.2 | 10 | 5860 | NP_001095869.1 | |
| NM_001297620.1 | 10 | 5903 | NP_001284549.1 | |
| NM_001297621.1 | 9 | 5674 | NP_001284550.1 | |
| NM_001297622.1 | 9 | 5737 | NP_001284551.1 | |
| NM_005826.4 | 11 | 6017 | NP_005817.1 | |
| AK001846.1 | 1 | 527 | ||
| BX647784.1 | 10 | 5758 |
More Information
Set Membership: |
|

Genomic Map