Assay Details
Target Gene Details
Entrez Gene ID: | 90806 |
Gene Name: | angel homolog 2 |
Gene Aliases: |
Ccr4d, KIAA0759L |
Location: |
Chr.1:212992182-213015875 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGEL2 | NM_001300753.1 | NP_001287682.1 | ||
| NM_001300755.1 | NP_001287684.1 | |||
| NM_001300757.1 | NP_001287686.1 | |||
| NM_001300758.1 | NP_001287687.1 | |||
| NM_144567.4 | NP_653168.2 | |||
| NR_125333.1 | ||||
| XM_005273344.1 | XP_005273401.1 | |||
| XM_005273345.1 | XP_005273402.1 | |||
| XM_005273346.2 | XP_005273403.1 | |||
| XM_005273347.3 | XP_005273404.1 | |||
| XM_017002774.1 | XP_016858263.1 | |||
| XM_017002775.1 | XP_016858264.1 | |||
| XM_017002776.1 | XP_016858265.1 | |||
| XM_017002777.1 | XP_016858266.1 | |||
| XM_017002778.1 | XP_016858267.1 | |||
| AF510741.1 | AAM44053.1 | |||
| AK093947.1 | ||||
| AK294479.1 | ||||
| AK295116.1 | ||||
| AK299208.1 | ||||
| AK299365.1 | ||||
| AK310946.1 | ||||
| AL079275.1 | ||||
| BC047469.1 | AAH47469.1 |
More Information
Set Membership: |
|

Genomic Map