Assay Details
Target Gene Details
Entrez Gene ID: | 26476 |
Gene Name: | olfactory receptor family 10 subfamily J member 1 |
Gene Aliases: |
HGMP07J, HSHGMP07J |
Location: |
Chr.1:159406348-159440967 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10J1 | XM_006711271.2 | XP_006711334.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78082752] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map