Assay Details
Target Gene Details
Entrez Gene ID: | 148753 |
Gene Name: | family with sequence similarity 163 member A |
Gene Aliases: |
C1orf76, NDSP |
Location: |
Chr.1:179727763-179816198 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM163A | XM_006711177.1 | XP_006711240.1 | ||
| XM_011509221.1 | XP_011507523.1 | |||
| XM_017000393.1 | XP_016855882.1 | |||
| AK292432.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548339 | Chr.1:179717109 - 179753692 on Build GRCh38 | Gain |
|
| dgv64e214 | Chr.1:179716874 - 179766076 on Build GRCh38 | Gain |
|

Genomic Map