Assay Details
Target Gene Details
Entrez Gene ID: | 3716 |
Gene Name: | Janus kinase 1 |
Gene Aliases: |
JAK1A, JAK1B, JTK3 |
Location: |
Chr.1:64833223-65067746 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAK1 | NM_001321852.1 | NP_001308781.1 | ||
| NM_001321853.1 | NP_001308782.1 | |||
| NM_001321854.1 | NP_001308783.1 |
Target Gene Details
Entrez Gene ID: | 101927084 |
Gene Name: | long intergenic non-protein coding RNA 1359 |
Gene Aliases: |
- |
Location: |
Chr.1:64979577-65002476 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01359 | NR_119383.1 | |||
| AK126239.1 | 1 | 457 | ||
| BI825893.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2676423 | Chr.1:64977641 - 65405798 on Build GRCh38 | Deletion |
|
| nsv830059 | Chr.1:64918047 - 65074478 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map