Assay Details
Target Gene Details
Entrez Gene ID: | 9698 |
Gene Name: | pumilio RNA binding family member 1 |
Gene Aliases: |
HSPUM, PUMH, PUMH1, PUML1 |
Location: |
Chr.1:30931506-31065717 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PUM1 | NM_001020658.1 | NP_001018494.1 | ||
| NM_014676.2 | NP_055491.1 | |||
| AF315592.1 | AAG31807.1 | |||
| AK056662.1 | ||||
| AK096476.1 | ||||
| AK222605.1 | BAD96325.1 | |||
| AK291779.1 | ||||
| AK294477.1 | ||||
| AK300457.1 | ||||
| BC013398.2 | AAH13398.1 | |||
| BX647437.1 | ||||
| D43951.2 | BAA07895.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545950 | Chr.1:30935209 - 31062316 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map