Assay Details
Target Gene Details
Entrez Gene ID: | 101929099 |
Gene Name: | uncharacterized LOC101929099 |
Gene Aliases: |
- |
Location: |
Chr.1:117025482-117059490 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929099 | NR_110786.1 | |||
| AK124329.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518833 | Chr.1:117055279 - 117059455 on Build GRCh38 | Loss |
|
| nsv1009706 | Chr.1:117049372 - 117442154 on Build GRCh38 | Gain |
|

Genomic Map