Assay Details
Target Gene Details
Entrez Gene ID: | 100506963 |
Gene Name: | TCEB3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:23760382-23778297 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCEB3-AS1 | NR_038280.1 | |||
| AL049675.1 | ||||
| BG429664.1 |

Genomic Map