Assay Details
Target Gene Details
Entrez Gene ID: | 80133 |
Gene Name: | maestro heat like repeat family member 9 |
Gene Aliases: |
ARMC11, C1orf129 |
Location: |
Chr.1:170935471-171066341 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MROH9 | NM_001163629.1 | NP_001157101.1 | ||
| NM_025063.2 | NP_079339.2 | |||
| XM_011510005.2 | XP_011508307.1 | |||
| XM_011510006.2 | XP_011508308.1 | |||
| AK027203.1 | BAB15690.1 | |||
| BC113390.1 | ||||
| BC113392.1 | ||||
| DB096780.1 | ||||
| DB211602.1 | ||||
| DB230154.1 |

Genomic Map