Assay Details
Target Gene Details
Entrez Gene ID: | 54361 |
Gene Name: | Wnt family member 4 |
Gene Aliases: |
SERKAL, WNT-4 |
Location: |
Chr.1:22117305-22143981 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WNT4 | NM_030761.4 | 5 | 3534 | NP_110388.2 |
| XM_011541597.2 | 5 | 5008 | XP_011539899.1 | |
| XM_011541598.2 | 5 | 3603 | XP_011539900.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6076 | Chr.1:22089806 - 22123911 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map