Assay Details
Target Gene Details
Entrez Gene ID: | 100302401 |
Gene Name: | RASAL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:178091508-178093993 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASAL2-AS1 | NR_027982.1 | 2 | 595 | |
| AK124464.1 | 2 | 229 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548252 | Chr.1:177758922 - 178428784 on Build GRCh38 | Gain |
|

Genomic Map