Assay Details
Target Gene Details
Entrez Gene ID: | 101928696 |
Gene Name: | uncharacterized LOC101928696 |
Gene Aliases: |
- |
Location: |
Chr.1:174934947-174954261 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928696 | NR_121196.1 | |||
| AI807953.1 | ||||
| BX102231.1 |
Target Gene Details
Entrez Gene ID: | 9910 |
Gene Name: | RAB GTPase activating protein 1 like |
Gene Aliases: |
HHL, TBC1D18 |
Location: |
Chr.1:174159414-174995308 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
More Information
Set Membership: |
|

Genomic Map