Assay Details
Target Gene Details
Entrez Gene ID: | 3953 |
Gene Name: | leptin receptor |
Gene Aliases: |
CD295, LEP-R, LEPRD, OB-R, OBR |
Location: |
Chr.1:65420652-65637493 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LEPR | NM_001003679.3 | NP_001003679.1 | ||
| NM_001003680.3 | NP_001003680.1 | |||
| NM_002303.5 | NP_002294.2 | |||
| AK290208.1 | ||||
| BC131779.1 | ||||
| U43168.1 | AAA93015.1 | |||
| U66495.1 | AAB07495.1 | |||
| U66496.1 | AAB07496.1 | |||
| U66497.1 | AAB07497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546438 | Chr.1:65396115 - 65484373 on Build GRCh38 | Gain |
|
| nsv830081 | Chr.1:65420397 - 65597117 on Build GRCh38 | Gain |
|
| nsv830070 | Chr.1:65334238 - 65516971 on Build GRCh38 | Gain |
|
| dgv174n100 | Chr.1:65386381 - 65506395 on Build GRCh38 | Gain |
|
| nsv528808 | Chr.1:65386076 - 65513234 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map