Assay Details
Target Gene Details
Entrez Gene ID: | 58 |
Gene Name: | actin, alpha 1, skeletal muscle |
Gene Aliases: |
ACTA, ASMA, CFTD, CFTD1, CFTDM, MPFD, NEM1, NEM2, NEM3, SHPM |
Location: |
Chr.1:229431245-229434096 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTA1 | NM_001100.3 | NP_001091.1 | ||
| AK096902.1 | ||||
| AK300697.1 | ||||
| AL598491.1 | ||||
| BC012597.1 | AAH12597.1 | |||
| BX648545.1 | ||||
| J00068.1 | AAB59376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv945360 | Chr.1:229431230 - 229435274 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map