Assay Details
Target Gene Details
Entrez Gene ID: | 9132 |
Gene Name: | potassium voltage-gated channel subfamily Q member 4 |
Gene Aliases: |
DFNA2, DFNA2A, KV7.4 |
Location: |
Chr.1:40784012-40840457 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ4 | NM_004700.3 | NP_004691.2 | ||
| NM_172163.2 | NP_751895.1 | |||
| AF105202.1 | AAD14680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577822 | Chr.1:40770274 - 40789804 on Build GRCh38 | Loss |
|
| nsv610 | Chr.1:40758199 - 40789878 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map