Assay Details
Target Gene Details
Entrez Gene ID: | 54583 |
Gene Name: | egl-9 family hypoxia inducible factor 1 |
Gene Aliases: |
C1orf12, ECYT3, HALAH, HIF-PH2, HIFPH2, HPH-2, HPH2, PHD2, SM20, ZMYND6 |
Location: |
Chr.1:231363751-231425044 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EGLN1 | NM_022051.2 | NP_071334.1 | ||
| XM_005273166.4 | XP_005273223.1 | |||
| XM_005273167.4 | XP_005273224.1 | |||
| AF334711.1 | ||||
| AI017372.1 | ||||
| AJ227859.1 | ||||
| AJ310543.1 | CAC42509.1 | |||
| AL833885.1 | CAD38741.2 |
More Information
Set Membership: |
|

Genomic Map