Assay Details
Target Gene Details
Entrez Gene ID: | 339751 |
Gene Name: | MLK7 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:173197712-173282036 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLK7-AS1 | NR_033882.1 |
Target Gene Details
Entrez Gene ID: | 51776 |
Gene Name: | sterile alpha motif and leucine zipper containing kinase AZK |
Gene Aliases: |
AZK, MLK7, MLT, MLTK, MRK, SFMMP, mlklak, pk |
Location: |
Chr.2:173075435-173268009 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZAK | NM_016653.2 | NP_057737.2 | ||
| NM_133646.2 | NP_598407.1 | |||
| XM_005246640.2 | XP_005246697.1 | |||
| XM_017004323.1 | XP_016859812.1 | |||
| XM_017004324.1 | XP_016859813.1 | |||
| AB030034.1 | BAB12040.1 | |||
| AB041883.1 | ||||
| AB049733.1 | BAB16444.1 | |||
| AB049734.1 | BAB16445.1 | |||
| AB208974.1 | BAD92211.1 | |||
| AF238255.1 | AAF63490.1 | |||
| AF251441.1 | AAF65822.1 | |||
| AF325454.1 | AAK11615.1 | |||
| AF465843.1 | AAO33376.1 | |||
| AF480461.1 | AAL85891.1 | |||
| AF480462.1 | AAL85892.1 | |||
| AK056310.1 | ||||
| AK291825.1 | ||||
| AK298634.1 | ||||
| BC001401.2 | AAH01401.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3037 | Chr.2:173190146 - 173223386 on Build GRCh38 | Insertion |
|
| nsv834460 | Chr.2:173054736 - 173220560 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77499294] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map