Assay Details
Target Gene Details
Entrez Gene ID: | 9167 |
Gene Name: | cytochrome c oxidase subunit 7A2 like |
Gene Aliases: |
COX7AR, COX7RP, EB1, SIG81 |
Location: |
Chr.2:42335561-42368957 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX7A2L | NM_001319036.1 | NP_001305965.1 | ||
| NM_001319037.1 | NP_001305966.1 | |||
| NM_001319038.1 | NP_001305967.1 | |||
| NM_001319040.1 | NP_001305969.1 | |||
| NM_004718.3 | NP_004709.2 | |||
| NR_134947.1 | ||||
| NR_134948.1 | ||||
| AB007618.1 | BAA22571.1 | |||
| AF127788.1 | AAF72746.1 | |||
| AK092911.1 | ||||
| AK130281.1 | ||||
| BC005251.1 | AAH05251.1 | |||
| BC007095.1 | AAH07095.1 | |||
| BF130603.1 | ||||
| BI598394.1 | ||||
| BT007371.1 | AAP36035.1 | |||
| CA336587.1 | ||||
| CD700827.1 | ||||
| CF593726.1 | ||||
| CR542208.1 | CAG47004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583758 | Chr.2:42347476 - 42362702 on Build GRCh38 | Loss |
|
| nsv524679 | Chr.2:42350030 - 42576225 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map