Assay Details
Target Gene Details
Entrez Gene ID: | 10096 |
Gene Name: | ARP3 actin related protein 3 homolog |
Gene Aliases: |
ARP3 |
Location: |
Chr.2:113889934-113961554 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR3 | NM_001277140.1 | NP_001264069.1 | ||
| NM_005721.4 | NP_005712.1 | |||
| NR_102318.1 | ||||
| AF006083.1 | AAB64188.1 | |||
| AF127773.1 | AAD51904.1 | |||
| AK300030.1 | ||||
| AK300224.1 | ||||
| AK302152.1 | ||||
| AK302359.1 | ||||
| AK303988.1 | ||||
| AK312659.1 | ||||
| AK316213.1 | ||||
| AK316396.1 | ||||
| BC044590.1 | AAH44590.1 | |||
| DB453995.1 |
More Information
Set Membership: |
|

Genomic Map