Assay Details
Target Gene Details
Entrez Gene ID: | 403150 |
Gene Name: | uncharacterized protein FLJ31356 |
Gene Aliases: |
- |
Location: |
Chr.2:28384409-28394672 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLJ31356 | NR_103831.1 | |||
| AK055918.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006830 | Chr.2:28331908 - 28419880 on Build GRCh38 | Gain |
|

Genomic Map