Assay Details
Target Gene Details
Entrez Gene ID: | 388962 |
Gene Name: | bolA family member 3 |
Gene Aliases: |
MMDS2 |
Location: |
Chr.2:74135401-74147912 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BOLA3 | NM_001035505.1 | NP_001030582.1 | ||
| NM_212552.2 | NP_997717.2 | |||
| AI652095.1 | ||||
| BC028282.1 | AAH28282.1 | |||
| DQ225187.1 | ABB04094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525117 | Chr.2:74131774 - 74147514 on Build GRCh38 | Loss |
|
| esv3591277 | Chr.2:74117985 - 74144281 on Build GRCh38 | Gain |
|
| esv3591278 | Chr.2:74130114 - 74143581 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map