Assay Details
Target Gene Details
Entrez Gene ID: | 6496 |
Gene Name: | SIX homeobox 3 |
Gene Aliases: |
HPE2 |
Location: |
Chr.2:44941898-44946077 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIX3 | NM_005413.3 | NP_005404.1 | ||
| AJ012611.1 | CAB42539.1 | |||
| BF569811.1 | ||||
| BF570321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526534 | Chr.2:44934159 - 44985100 on Build GRCh38 | Loss |
|
| nsv953779 | Chr.2:44932862 - 44955461 on Build GRCh38 | Deletion |
|
| nsv457207 | Chr.2:44931099 - 44967150 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map