Assay Details
Target Gene Details
Entrez Gene ID: | 3557 |
Gene Name: | interleukin 1 receptor antagonist |
Gene Aliases: |
DIRA, ICIL-1RA, IL-1RN, IL-1ra, IL-1ra3, IL1F3, IL1RA, IRAP, MVCD4 |
Location: |
Chr.2:113099365-113134016 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1RN | XM_011511121.1 | XP_011509423.1 |
More Information
Set Membership: |
|

Genomic Map