Assay Details
Target Gene Details
Entrez Gene ID: | 431705 |
Gene Name: | astacin-like metallo-endopeptidase (M12 family) |
Gene Aliases: |
SAS1B |
Location: |
Chr.2:96122817-96143059 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASTL | XM_011511206.2 | XP_011509508.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion |
|
| dgv4009n100 | Chr.2:96072837 - 96159625 on Build GRCh38 | Gain |
|
| nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion |
|
| esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map