Assay Details
Target Gene Details
Entrez Gene ID: | 220988 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A3 |
Gene Aliases: |
2610510D13Rik, D10S102, FBRNP, HNRPA3 |
Location: |
Chr.2:177212694-177223959 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPA3 | NM_194247.2 | NP_919223.1 | ||
| XM_005246380.1 | XP_005246437.1 | |||
| XM_005246381.4 | XP_005246438.1 | |||
| XM_006712365.2 | XP_006712428.1 | |||
| XM_017003587.1 | XP_016859076.1 | |||
| XM_017003588.1 | XP_016859077.1 | |||
| AK293125.1 | ||||
| AK303208.1 | ||||
| AK304687.1 | ||||
| AK316434.1 | ||||
| AY363225.1 | AAQ63629.1 | |||
| BC112027.1 | AAI12028.1 | |||
| BC113470.1 | ||||
| BX647217.1 | ||||
| DB093606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834465 | Chr.2:177202566 - 177250385 on Build GRCh38 | Loss |
|
| nsv963751 | Chr.2:177214885 - 177221027 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map