Assay Details
Target Gene Details
Entrez Gene ID: | 151306 |
Gene Name: | G protein-coupled bile acid receptor 1 |
Gene Aliases: |
BG37, GPCR19, GPR131, M-BAR, TGR5 |
Location: |
Chr.2:218259496-218263861 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPBAR1 | NM_001077191.1 | NP_001070659.1 | ||
| NM_001077194.1 | NP_001070662.1 | |||
| NM_001321950.1 | NP_001308879.1 | |||
| NM_170699.2 | NP_733800.1 | |||
| XM_011510743.1 | XP_011509045.1 | |||
| XM_017003467.1 | XP_016858956.1 | |||
| XM_017003468.1 | XP_016858957.1 | |||
| XM_017003469.1 | XP_016858958.1 | |||
| AK122658.1 | ||||
| AK122660.1 | ||||
| BC033625.1 | AAH33625.1 | |||
| BI834364.1 |
More Information
Set Membership: |
|

Genomic Map