Assay Details
Target Gene Details
Entrez Gene ID: | 3431 |
Gene Name: | SP110 nuclear body protein |
Gene Aliases: |
IFI41, IFI75, IPR1, VODI |
Location: |
Chr.2:230165466-230225729 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998291 | Chr.2:230180672 - 230271563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map