Assay Details
Target Gene Details
Entrez Gene ID: | 200558 |
Gene Name: | aprataxin and PNKP like factor |
Gene Aliases: |
APFL, C2orf13, PALF, Xip1, ZCCHH1 |
Location: |
Chr.2:68467559-68580162 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APLF | NM_173545.2 | NP_775816.1 | ||
| AK290841.1 | ||||
| AK308186.1 | ||||
| BC041144.1 | AAH41144.1 | |||
| DA641644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3591168 | Chr.2:68463983 - 68522612 on Build GRCh38 | Loss |
|
| esv3892603 | Chr.2:68459705 - 68514828 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map