Assay Details
Target Gene Details
Entrez Gene ID: | 100169989 |
Gene Name: | diazepam binding inhibitor-like 5 pseudogene 2 |
Gene Aliases: |
ELP1P |
Location: |
Chr.2:63117851-63119542 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DBIL5P2 | NR_036635.2 | 1 | 631 | |
| AF229803.1 | 1 | 631 | AAF79121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004678 | Chr.2:63003587 - 63335349 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map