Assay Details
Target Gene Details
Entrez Gene ID: | 9654 |
Gene Name: | tubulin tyrosine ligase like 4 |
Gene Aliases: |
- |
Location: |
Chr.2:218710835-218757484 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL4 | NM_014640.4 | NP_055455.3 | ||
| XM_006712875.2 | XP_006712938.1 | |||
| XM_017005387.1 | XP_016860876.1 | |||
| XM_017005388.1 | XP_016860877.1 | |||
| XM_017005389.1 | XP_016860878.1 | |||
| XM_017005390.1 | XP_016860879.1 | |||
| XM_017005391.1 | XP_016860880.1 | |||
| AK291770.1 | ||||
| AK302728.1 | ||||
| AL833511.1 | ||||
| BC021707.2 | AAH21707.1 | |||
| D79995.1 | ||||
| DB097392.1 |
More Information
Set Membership: |
|

Genomic Map