Assay Details
Target Gene Details
Entrez Gene ID: | 1385 |
Gene Name: | cAMP responsive element binding protein 1 |
Gene Aliases: |
CREB, CREB-1 |
Location: |
Chr.2:207529892-207605989 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB1 | NM_001320793.1 | NP_001307722.1 | ||
| NM_004379.4 | NP_004370.1 | |||
| NM_134442.4 | NP_604391.1 | |||
| NR_135473.1 | ||||
| XM_011510645.1 | XP_011508947.1 | |||
| XM_011510646.2 | XP_011508948.1 | |||
| XM_011510647.2 | XP_011508949.1 | |||
| XM_011510648.2 | XP_011508950.1 | |||
| XM_011510649.2 | XP_011508951.1 | |||
| XM_017003399.1 | XP_016858888.1 | |||
| XM_017003400.1 | XP_016858889.1 | |||
| AK298740.1 | ||||
| AK312747.1 | ||||
| AK315226.1 | ||||
| AY347527.1 | AAQ24858.1 | |||
| BC010636.1 | AAH10636.1 | |||
| BC095407.1 | AAH95407.1 | |||
| DC366797.1 | ||||
| M27691.1 | AAA35715.1 |
More Information
Set Membership: |
|

Genomic Map